Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009204.2 | NP_033230.2 | 3, 4 | 540 | ||
GenBank mRNA | AB008453.1 | - | 3, 4 | 338 | ||
AK137607.1 | - | 3, 4 | 525 | |||
AK138832.1 | - | 3, 4 | 514 | |||
AK160882.1 | - | 3, 4 | 526 | |||
AK164310.1 | - | 3, 4 | 515 | |||
BC014282.1 | - | 3, 4 | 510 | 4207674 | ||
M23383.1 | - | 3, 4 | 490 |