Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009208.3 | NP_033234.2 | 4 | 544 | ||
GenBank mRNA | AK045583.1 | - | 3 | 431 | ||
AK147218.1 | - | 556 | ||||
BC066086.1 | - | 4 | 430 | |||
BC080271.1 | - | 4 | 539 | 6831942 | ||
M28383.1 | - | 4 | 514 |