Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168388.2 | NP_001161860.1 | 2 | 390 | ||
NM_001168389.2 | NP_001161861.2 | 2 | 373 | |||
NM_006079.4 | NP_006070.2 | 2 | 529 | |||
GenBank mRNA | AF109161.1 | - | 2 | 499 | ||
BC004377.1 | - | 2 | 482 | 3640855 | ||
BG900380.1 | - | 2 | 427 | |||
BI763038.1 | - | 2 | 373 | |||
BM763150.1 | - | 2 | 527 | |||
U65093.1 | - | 2 | 482 |