Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006694.3 | NP_006685.1 | 2, 3 | 836 | ||
GenBank mRNA | AB016488.1 | - | 2, 3 | 545 | ||
AF115850.2 | - | 2, 3 | 534 | |||
AF131797.1 | - | 2, 3 | 417 | |||
AK312106.1 | - | 2, 3 | 518 | |||
BC000499.1 | - | 2, 3 | 558 | 2820861 | ||
BC000996.2 | - | 2, 3 | 485 | 2901221 | ||
BC001363.1 | - | 2, 3 | 523 | 3050800 | ||
BC001667.1 | - | 2, 3 | 488 | 2823348 | ||
BC019277.1 | - | 2, 3 | 479 | 2823348 | ||
BM907938.1 | - | 597 | ||||
BT007285.1 | - | 2, 3 | 113 | |||
CR456985.1 | - | 2, 3 | 113 |