Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009162.3 | NP_033188.3 | 6 | 735 | ||
GenBank mRNA | AK003049.1 | - | 6 | 677 | ||
AK005331.1 | - | 6 | 684 | |||
AK011938.1 | - | 6 | 666 | |||
AK017481.1 | - | 6 | 788 | |||
AK019337.1 | - | 6 | 736 | |||
AK031856.1 | - | 6 | 669 | |||
AK161556.1 | - | 6 | 674 | |||
AK161579.1 | - | 6 | 676 | |||
BC029021.1 | - | 6 | 717 | 5361810 | ||
X15830.1 | - | 6 | 713 |