Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034962.1 | NP_001030134.1 | 23 | 2252 | ||
NM_001034963.1 | NP_001030135.1 | 23 | 2282 | |||
NM_001034964.1 | NP_001030136.1 | 22 | 2245 | |||
NM_009166.3 | NP_033192.2 | 24 | 2441 | |||
NM_178362.1 | NP_848139.1 | 29 | 3290 | |||
GenBank mRNA | AF078666.1 | - | 23 | 2271 | ||
AF078667.1 | - | 23 | 2241 | |||
AF521593.1 | - | 21 | 2202 | |||
AK087553.1 | - | 29 | 3292 | |||
AK122487.2 | - | 27 | 4014 | |||
AK146712.1 | - | 21 | 2079 | |||
AK160958.1 | - | 1 | 26 | |||
BC012703.1 | - | 24 | 2274 | 3602997 | ||
U58883.1 | - | 24 | 2379 |