Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009191.3 | NP_033217.1 | 16 | 2208 | ||
GenBank mRNA | AK138593.1 | - | 16 | 2246 | ||
AK141383.1 | - | 16 | 2217 | |||
AK147772.1 | - | 16 | 1929 | |||
AK161996.1 | - | 109 | ||||
BC048175.1 | - | 16 | 2208 | 6825472 |