Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289724.1 | NP_001276653.1 | 6, 7 | 971 | ||
NM_001289725.1 | NP_001276654.1 | 6, 7 | 926 | |||
NM_009193.2 | NP_033219.1 | 7, 8 | 1031 | |||
GenBank mRNA | AK136165.1 | - | 3, 4 | 321 | ||
AK154053.1 | - | 7, 8 | 778 | |||
AK162121.1 | - | 7, 8 | 777 | |||
BC025113.1 | - | 7, 8 | 765 | |||
BC029631.1 | - | 6, 7 | 703 | 3500676 | ||
U75680.1 | - | 7, 8 | 763 |