Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170638.1 | NP_001164109.1 | 13 | 1888 | ||
NM_009198.3 | NP_033224.3 | 13 | 1980 | |||
GenBank mRNA | AK132800.1 | - | 13 | 2459 | ||
AK164712.1 | - | 13 | 1890 | |||
BC013445.1 | - | 13 | 1921 | 4241557 | ||
X77241.1 | - | 13 | 1847 |