Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009203.3 | NP_033229.3 | 10 | 1937 | ||
GenBank mRNA | AB005451.1 | - | 10 | 1718 | ||
AK142580.1 | - | 9 | 1086 | |||
BC035927.1 | - | 1873 | 4242354 |