Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009204.2 | NP_033230.2 | 11 | 2258 | ||
GenBank mRNA | AK137607.1 | - | 11 | 2243 | ||
AK160882.1 | - | 9 | 2883 | |||
AK164310.1 | - | 11 | 2233 | |||
BC014282.1 | - | 11 | 2228 | 4207674 | ||
M23383.1 | - | 11 | 2204 |