Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 3 | 444 | ||
GenBank mRNA | AK042194.1 | - | 4 | 357 | ||
AK133922.1 | - | 3 | 419 | |||
AK143485.1 | - | 3 | 432 | |||
AK144122.1 | - | 3 | 418 | |||
AK144631.1 | - | 3 | 412 | |||
AK145352.1 | - | 3 | 411 | |||
AK183841.1 | - | 3 | 218 | |||
AK185018.1 | - | 3 | 219 | |||
BC022173.1 | - | 3 | 408 | |||
BC052423.1 | - | 3 | 406 | 6400646 | ||
BC053064.1 | - | 3 | 400 | |||
BC056375.1 | - | 3 | 407 | |||
U85614.1 | - | 3 | 405 |