Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009215.1 | NP_033241.1 | 1, 2 | 246 | ||
GenBank mRNA | AK003014.1 | - | 1, 2 | 247 | ||
BC010770.1 | - | 1, 2 | 236 | 4218815 |