Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005045.2 | - | 11 | 2808 | ||
AK077425.1 | - | 10 | 3833 | |||
AK133115.1 | - | 10 | 1449 | |||
AK146729.1 | - | 10 | 1449 | |||
AK148013.1 | - | 10 | 1477 | |||
AK180798.1 | - | 1 | 31 | |||
BC002169.1 | - | 4 | 627 | |||
BC049128.1 | - | 10 | 3780 | |||
BC058513.1 | - | 3 | 954 | |||
BC094067.1 | - | 3 | 1105 | |||
BC131994.1 | - | 10 | 1341 | 40130271 | ||
BC132020.1 | - | 10 | 1341 | 40130297 |