Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009260.2 | NP_033286.2 | 4 | 874 | ||
NM_175836.2 | NP_787030.2 | 4 | 534 | |||
GenBank mRNA | AF017112.1 | - | 4 | 781 | ||
AK079842.1 | - | 4 | 864 | |||
AK157727.1 | - | 4 | 864 | |||
AK169544.1 | - | 4 | 874 | |||
AK220456.1 | - | 5 | 736 | |||
M74773.1 | - | 4 | 854 |