Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009284.2 | NP_033310.2 | 3 | 512 | ||
GenBank mRNA | AF481808.1 | - | 2 | 227 | ||
AF481809.1 | - | 2 | 227 | |||
AK149563.1 | - | 3 | 472 | |||
AK163053.1 | - | 3 | 513 | |||
AK217738.1 | - | 2 | 255 | |||
BC127054.1 | - | 3 | 302 | 40109712 |