Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syngr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009304.2 | NP_033330.1 | 2 | 184 | ||
GenBank mRNA | AF151985.1 | - | 2 | 181 | ||
AF364049.1 | - | 2 | 164 | |||
AJ002307.1 | - | 2 | 150 | |||
AK150234.1 | - | 2 | 184 | |||
AK157343.1 | - | 2 | 156 | |||
AK170681.1 | - | 2 | 184 | |||
BC004829.1 | - | 2 | 177 | 2646657 | ||
BY146297.1 | - | 2 | 185 |