Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009307.3 | NP_033333.2 | 3 | 542 | ||
GenBank mRNA | AB036514.1 | - | 2 | 337 | ||
AF257304.1 | - | 2 | 297 | |||
AK036357.1 | - | 3 | 543 | |||
BC027019.1 | - | 2 | 303 | 5362352 | ||
D37793.1 | - | 1 | 176 |