Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009225.2 | NP_033251.1 | 3 | 386 | ||
GenBank mRNA | AK077537.1 | - | 3 | 339 | ||
AK146319.1 | - | 3 | 429 | |||
AK178243.1 | - | 2 | 186 | |||
AK186005.1 | - | 2 | 186 | |||
AK203845.1 | - | 2 | 186 | |||
AK207139.1 | - | 2 | 186 | |||
BG086527.2 | - | 3 | 344 |