Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009253.2 | NP_033279.2 | 1203 | |||
GenBank mRNA | BC011158.1 | - | 1205 | 4194074 | ||
BC053337.1 | - | 1181 | 4208766 | |||
X55148.1 | - | 1175 | ||||
X69832.1 | - | 699 |