Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009257.3 | NP_033283.1 | 2 | 232 | ||
GenBank mRNA | AK136532.1 | - | 2 | 238 | ||
AK145220.1 | - | 1 | 200 | |||
BC005434.1 | - | 2 | 233 | 3600282 | ||
U54705.1 | - | 3 | 339 |