Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009275.4 | NP_033301.1 | 4 | 382 | ||
GenBank mRNA | AK004854.1 | - | 4 | 415 | ||
AK007859.1 | - | 4 | 388 | |||
AK008383.1 | - | 4 | 425 | |||
AK015469.1 | - | 4 | 447 | |||
AK030437.1 | - | 4 | 383 | |||
AK075583.1 | - | 4 | 385 | |||
AK160600.1 | - | 3 | 1616 | |||
AK167039.1 | - | 4 | 388 | |||
BC003798.1 | - | 4 | 403 | 3490766 | ||
U17343.1 | - | 4 | 396 |