Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009282.3 | NP_033308.2 | 3 | 345 | ||
GenBank mRNA | AK017978.1 | - | 3 | 333 | ||
AK043215.1 | - | 3 | 521 | |||
AK048671.1 | - | 3 | 521 | |||
AK052003.1 | - | 3 | 353 | |||
AK134316.1 | - | 3 | 370 | |||
BC062954.1 | - | 3 | 345 | 30537388 | ||
BC069841.1 | - | 3 | 200 | |||
Z75332.1 | - | 3 | 394 |