Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009305.2 | NP_033331.2 | 4, 5 | 536 | ||
GenBank mRNA | AK034291.1 | - | 4, 5 | 479 | ||
AK043756.1 | - | 4, 5 | 433 | |||
AK090259.1 | - | 4, 5 | 434 | |||
BC014823.1 | - | 4, 5 | 406 | 4507116 | ||
BC052354.1 | - | 4, 5 | 430 | |||
BC115644.1 | - | 3, 4 | 389 | |||
BC115645.1 | - | 3, 4 | 388 | |||
X95818.1 | - | 4, 5 | 405 |