Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009320.4 | NP_033346.2 | 6, 7 | 1037 | ||
GenBank mRNA | AK143510.1 | - | 6, 7 | 1001 | ||
BC015245.1 | - | 6, 7 | 974 | 4242193 | ||
L03292.2 | - | 6, 7 | 1009 |