Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009214.3 | NP_033240.3 | 11 | 1533 | ||
GenBank mRNA | AF031486.1 | - | 11 | 1516 | ||
AK002687.1 | - | 11 | 1530 | |||
AK017775.1 | - | 11 | 1504 | |||
AK049787.1 | - | 11 | 1522 | |||
AK136798.1 | - | 11 | 1536 | |||
AK160659.1 | - | 11 | 1535 | |||
AK166665.1 | - | 11 | 1559 | |||
BC046623.1 | - | 11 | 1515 | 6315420 | ||
BC058688.1 | - | 11 | 1774 | 6413869 | ||
Y09419.1 | - | 11 | 1506 |