Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sstr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009219.3 | NP_033245.2 | 1 | 585 | ||
GenBank mRNA | AK051189.1 | - | 620 | |||
BC138487.1 | - | 564 | 8861507 | |||
BC138488.1 | - | 564 | 8861508 |
Set Membership: |
G-protein coupled receptor |