Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166385.2 | NP_001159857.2 | 5, 6 | 2310 | ||
NM_009239.4 | NP_033265.4 | 5, 6 | 2316 | |||
GenBank mRNA | AB221633.1 | - | 4, 5 | 2109 | ||
BC076630.1 | - | 5, 6 | 2240 | 30637762 | ||
U62522.1 | - | 5, 6 | 2245 |