Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009257.3 | NP_033283.1 | 7 | 2204 | ||
GenBank mRNA | AK004131.1 | - | 1 | 99 | ||
AK145220.1 | - | 6 | 2171 | |||
BC005434.1 | - | 7 | 2205 | 3600282 |