Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009260.2 | NP_033286.2 | 27 | 6257 | ||
NM_175836.2 | NP_787030.2 | 27 | 5917 | |||
GenBank mRNA | AF017112.1 | - | 27 | 6164 | ||
AK051630.1 | - | 2 | 275 | |||
AK161119.1 | - | 6 | 1040 | |||
AK220456.1 | - | 28 | 6119 | |||
M74773.1 | - | 27 | 6237 |