Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 3 | 452 | ||
XM_006715880.3 | XP_006715943.1 | 3 | 425 | |||
GenBank mRNA | AK290351.1 | - | 3 | 416 | ||
AK313263.1 | - | 3 | 425 | |||
AU131228.1 | - | 3 | 411 | |||
BC130291.1 | - | 3 | 436 | 40146322 | ||
BC144156.1 | - | 3 | 436 | |||
CN289899.1 | - | 1 | 128 | |||
EU826604.1 | - | 3 | 365 | |||
EU826605.1 | - | 3 | 365 | |||
M18391.1 | - | 3 | 458 |