Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009275.4 | NP_033301.1 | 7 | 2979 | ||
GenBank mRNA | AK004854.1 | - | 7 | 3061 | ||
AK030437.1 | - | 7 | 2980 | |||
AK167039.1 | - | 7 | 2986 |