Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009284.2 | NP_033310.2 | 22 | 3205 | ||
GenBank mRNA | AK135911.1 | - | 5 | 968 | ||
AK149563.1 | - | 22 | 3166 | |||
AK163053.1 | - | 22 | 3206 | |||
AK180048.1 | - | 1 | 240 | |||
BC029318.1 | - | 9 | 1394 |