Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009288.2 | NP_033314.2 | 19 | 4968 | ||
GenBank mRNA | AA182088.1 | - | 1 | 348 | ||
AK148118.1 | - | 19 | 4929 | |||
AK220448.1 | - | 19 | 4866 | |||
BC069840.1 | - | 19 | 5037 | |||
BC096624.1 | - | 19 | 5049 | |||
D89728.1 | - | 19 | 4905 |