Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Surf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009298.3 | NP_033324.1 | 5 | 2256 | ||
GenBank mRNA | AK075636.2 | - | 1 | 1360 | ||
AK077989.1 | - | 5 | 2317 | |||
AK132194.1 | - | 5 | 2293 | |||
BC026514.1 | - | 2 | 1705 | |||
BC037634.1 | - | 5 | 2303 | 4952465 | ||
X92842.1 | - | 5 | 2241 |