Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003889.3 | NP_003880.3 | 2 | 1853 | ||
NM_022002.2 | NP_071285.1 | 2 | 179 | |||
NM_033013.2 | NP_148934.1 | 2 | 1853 | |||
GenBank mRNA | AB307701.1 | - | 1 | 14 | ||
AF061056.1 | - | 2 | 317 | |||
AF084644.1 | - | 2 | 190 | |||
AF084645.1 | - | 2 | 293 | |||
AJ009936.1 | - | 3 | 1853 | |||
AJ009937.1 | - | 3 | 1853 | |||
AK223456.1 | - | 2 | 298 | |||
AK310789.1 | - | 2 | 302 | |||
AY091855.1 | - | 2 | 596 | |||
BC017304.2 | - | 2 | 253 | 5087401 | ||
HQ692837.1 | - | 2 | 131 | |||
HQ709177.1 | - | 2 | 131 |