Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009369.4 | NP_033395.1 | 2 | 211 | ||
GenBank mRNA | AK084431.1 | - | 2 | 229 | ||
AK135536.1 | - | 2 | 256 | |||
AK142323.1 | - | 2 | 231 | |||
AK150449.1 | - | 2 | 229 | |||
AK150833.1 | - | 2 | 229 | |||
AK150969.1 | - | 2 | 230 | |||
AK151680.1 | - | 2 | 226 | |||
AK152365.1 | - | 2 | 230 | |||
AK152662.1 | - | 2 | 229 | |||
AK155572.1 | - | 2 | 226 | |||
AK155828.1 | - | 2 | 226 | |||
AK159711.1 | - | 2 | 226 | |||
AK164373.1 | - | 2 | 229 | |||
AK169912.1 | - | 2 | 231 | |||
AK170495.1 | - | 2 | 226 | |||
AK171557.1 | - | 1 | 50 | |||
BC129900.1 | - | 2 | 204 | 40111549 | ||
BC129901.1 | - | 2 | 204 | 40111550 | ||
L19932.1 | - | 2 | 212 |