Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164074.1 | NP_001157546.1 | 2 | 209 | ||
NM_001164075.1 | NP_001157547.1 | 2 | 549 | |||
NM_001164076.1 | NP_001157548.1 | 2 | 275 | |||
NM_001164077.1 | NP_001157549.1 | 1 | 503 | |||
NM_009372.3 | NP_033398.2 | 2 | 463 | |||
GenBank mRNA | AK008909.1 | - | 2 | 465 | ||
AK160193.1 | - | 2 | 168 | |||
BC005724.1 | - | 2 | 462 | 3595273 | ||
BC012700.1 | - | 2 | 560 | 4011149 | ||
BY096128.1 | - | 2 | 213 | |||
CD563532.1 | - | 2 | 379 | |||
X89749.1 | - | 2 | 419 |