Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009376.2 | NP_033402.2 | 2, 3 | 221 | ||
GenBank mRNA | AK135110.1 | - | 2, 3 | 216 | ||
AK160633.1 | - | 2, 3 | 230 | |||
AK168395.1 | - | 2, 3 | 187 | |||
BC012250.1 | - | 2, 3 | 201 | 4019334 | ||
L31959.1 | - | 2, 3 | 166 |