Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009390.2 | NP_033416.2 | 1 | 744 | ||
GenBank mRNA | AK080919.1 | - | 1 | 760 | ||
AK082541.1 | - | 2 | 798 | |||
U34042.1 | - | 1 | 744 |