Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009391.3 | NP_033417.1 | 4 | 311 | ||
GenBank mRNA | AF159256.1 | - | 3 | 177 | ||
AK002436.1 | - | 4 | 401 | |||
AK010569.1 | - | 3 | 243 | |||
AK010672.1 | - | 4 | 220 | |||
AK012268.1 | - | 3 | 280 | |||
AK075900.1 | - | 4 | 249 | |||
AK087974.1 | - | 4 | 222 | |||
AK145272.1 | - | 4 | 250 | |||
AK151341.1 | - | 4 | 243 | |||
AK151937.1 | - | 4 | 243 | |||
AK153197.1 | - | 4 | 257 | |||
AK153411.1 | - | 4 | 243 | |||
AK160647.1 | - | 4 | 250 | |||
AK167040.1 | - | 4 | 222 | |||
AK168183.1 | - | 4 | 249 | |||
BC014829.1 | - | 4 | 258 | 3968440 | ||
BC083356.1 | - | 4 | 215 | 6818977 | ||
L32751.1 | - | 3 | 202 | |||
S83456.1 | - | 4 | 288 |