Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002848.3 | NP_002839.1 | 13 | 2716 | ||
NM_030667.2 | NP_109592.1 | 13 | 2716 | |||
NM_030668.2 | NP_109593.1 | 1 | 307 | |||
NM_030669.2 | NP_109594.1 | 1 | 307 | |||
NM_030670.2 | NP_109595.1 | 1 | 307 | |||
NM_030671.2 | NP_109596.1 | 1 | 307 | |||
XM_017019725.1 | XP_016875214.1 | 13 | 2688 | |||
GenBank mRNA | AF152378.1 | - | 1 | 299 | ||
AF187042.1 | - | 1 | 299 | |||
AF187043.1 | - | 1 | 299 | |||
AF187044.1 | - | 1 | 299 | |||
AK289784.1 | - | 13 | 2461 | |||
AK290982.1 | - | 1 | 30 | |||
BC126201.1 | - | 13 | 2416 | 8991917 | ||
BC126203.1 | - | 13 | 2416 | 8991919 | ||
DC358516.1 | - | 1 | 308 | |||
U20489.1 | - | 13 | 2418 | |||
Z48541.1 | - | 13 | 2366 |