Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009403.3 | NP_033429.1 | 1, 2 | 443 | ||
GenBank mRNA | AK030916.1 | - | 1, 2 | 444 | ||
AK157270.1 | - | 1, 2 | 196 | |||
BC117093.1 | - | 1, 2 | 382 | 40126412 | ||
BC117097.1 | - | 1, 2 | 382 | 40126416 | ||
L09754.1 | - | 1, 2 | 330 |