Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303043.1 | NP_001289972.1 | 2 | 1364 | ||
NM_030935.4 | NP_112197.1 | 2 | 1467 | |||
NR_130118.1 | 2 | 622 | ||||
NR_130119.1 | 2 | 619 | ||||
GenBank mRNA | AJ133115.1 | - | 2 | 1052 | ||
AK296580.1 | - | 2 | 915 | |||
AK297702.1 | - | 2 | 557 | |||
AK314824.1 | - | 2 | 1262 | |||
BC001486.2 | - | 2 | 1467 | 2966929 | ||
BC001966.1 | - | 2 | 1320 | 3457297 | ||
BC002972.1 | - | 2 | 1374 | 3538175 | ||
BC010406.2 | - | 2 | 1381 | 3832780 | ||
BC031622.1 | - | 2 | 1357 | 5174360 | ||
BX341171.2 | - | 2 | 571 |