Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166584.1 | NP_001160056.1 | 2, 3 | 659 | ||
NM_001166585.1 | NP_001160057.1 | 1, 2 | 252 | |||
NM_009346.3 | NP_033372.1 | 2, 3 | 659 | |||
GenBank mRNA | AK140307.1 | - | 2, 3 | 660 | ||
AK148375.1 | - | 2, 3 | 625 | |||
BC052021.1 | - | 1, 2 | 193 | |||
BC060138.1 | - | 1, 2 | 193 | 6410573 | ||
L06865.1 | - | 1, 2 | 198 | |||
L13853.1 | - | 2, 3 | 386 | |||
S74227.1 | - | 2, 3 | 659 |