Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009357.2 | NP_033383.1 | 6 | 1036 | ||
GenBank mRNA | AK149789.1 | - | 6 | 1001 | ||
AK157834.1 | - | 4 | 1850 | |||
BC003802.1 | - | 6 | 980 | 3486968 | ||
X81058.1 | - | 6 | 983 |