Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 15 | 1807 | ||
GenBank mRNA | AB053464.1 | - | 15 | 1752 | ||
AF316616.2 | - | 15 | 1664 | |||
AK154050.1 | - | 15 | 1783 | |||
AK160587.1 | - | 15 | 1748 | |||
AK178339.1 | - | 2 | 149 | |||
AY338234.1 | - | 15 | 1737 | |||
BC009163.1 | - | 4 | 468 | |||
BC010716.1 | - | 15 | 1720 | 2655861 | ||
X81059.1 | - | 4 | 247 |