Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IAPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000415.2 | NP_000406.1 | 3 | 378 | ||
XM_017019253.1 | XP_016874742.1 | 3 | 327 | |||
GenBank mRNA | AA446156.1 | - | 3 | 294 | ||
CK905016.1 | - | 3 | 402 | |||
DQ516082.1 | - | 2 | 226 | |||
J04422.1 | - | 3 | 362 | |||
X14904.1 | - | 3 | 324 | |||
X14905.1 | - | 1 | 14 |