Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009407.2 | NP_033433.1 | 2 | 259 | ||
GenBank mRNA | AK006495.1 | - | 2 | 253 | ||
BC048494.1 | - | 2 | 259 | 6775189 | ||
BC061170.1 | - | 2 | 252 | 6773623 | ||
X12521.1 | - | 2 | 232 |