Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009428.2 | NP_033454.1 | 2 | 573 | ||
GenBank mRNA | AF029983.1 | - | 2 | 228 | ||
AF060107.1 | - | 2 | 572 | |||
AJ006204.1 | - | 1 | 170 | |||
AK038531.1 | - | 2 | 397 | |||
BC112972.1 | - | 2 | 391 | 40049027 |